TX-200 Rotor Tube Adapters
TX-200 Rotor Tube Adapters
Actual product may vary
TX-200 Rotor Tube Adapters
TX-200 Rotor Tube Adapters
TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500380810 mL Blood Collection or 15 mL Corex /Kimble Tube
7500377115 mL Conical Tube
750037855/7 mL Blood Collection Tube
7500380350 mL Conical Tube
Catalog number 75003808
Price (MYR)
-
Capacity:
10 mL Blood Collection or 15 mL Corex /Kimble Tube
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
Specifications
TypeRotor Tube Adapter
Capacity10 mL Blood Collection or 15 mL Corex /Kimble Tube
Max. Tube Diameter17 mm
Max. Tube Length121 mm
Unit SizeEA