8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges)
8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges)
Actual product may vary
8 x 50 Individually Sealed Fixed Angle Rotor for 50 mL conical (for compact and 1.6L General Purpose Centrifuges)
Centrifuge eight conical tubes in individually sealed vessels.
Have Questions?
Catalog NumberCapacity
750036948 x 50 mL
Catalog number 75003694
Price (MYR)
-
Capacity:
8 x 50 mL
Centrifuge eight conical tubes in individually sealed vessels using Thermo Scientific™ 8 x 50mL Sealed Fixed-Angle Rotor.
Individual sealed canisters for high-risk samples, certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Specifications
Description8 x 50mL Individually Sealed Fixed Angle Rotor
TypeFixed Angle Rotor
Capacity8 x 50 mL
Max. RCF6,755 x g
Max. Speed6,500 rpm
Radius (Metric)143 mm
Max. Tube Length116 mm
Max. Tube Diameter29.5 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Angle25°
Unit SizeEA