Fiberlite™ F15-8 x 50cy Fixed Angle Rotor
Fiberlite™ F15-8 x 50cy Fixed Angle Rotor
Actual product may vary
Thermo Scientific
Fiberlite™ F15-8 x 50cy Fixed Angle Rotor
Pellet fractional samples with high RCF using this fixed angle rotor.
Have Questions?
Catalog NumberMax. SpeedMax. RCFFor Use With (Equipment)
7500366314,500 rpm24,446 x gThermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Multifuge X3/X3F, Multifuge X1 and SL 40F Centrifuges
Catalog number 75003663
Price (MYR)
-
Max. Speed:
14,500 rpm
Max. RCF:
24,446 x g
For Use With (Equipment):
Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Multifuge X3/X3F, Multifuge X1 and SL 40F Centrifuges
Pellet fractional samples with high RCF using the Thermo ScientificTM FiberliteTM F15-8 x 50cy Fixed-Angle Rotor.
Fiberlite Rotor:
  • Reduces run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensures structural integrity through corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering Information:
Adapters are sold separately. O-ring Refresher Kit (021-085-077) is also available. Contact your sales representative for more information.
Compatible with:
For Thermo Scientific Sorvall™ Legend™ X1, XT, Heraeus™ Multifuge™ X1, X3, and Thermo Scientific Nalgene™ bottles and tubes
Recommended for:
Pelleting cells, bacteria, subcellular fractions, and small amounts of nucleic acids and viruses.
Note:
Performance may vary by centrifuge model.
Specifications
DescriptionFiberlite F15-8 x 50cy Fixed Angle Rotor for Thermo Scientific Sorvall Legend X1, Sorvall Legend XT
TypeFixed Angle Rotor
Capacity8 x 50 mL
Max. RCF24,446 x g
Max. Speed14,500 rpm
Radius (Metric)104 mm
K Factor1381
Max. Tube Length121 mm
Max. Tube Diameter30 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Multifuge X3/X3F, Multifuge X1 and SL 40F Centrifuges
Angle25°
BiocontainmentYes
Green FeaturesEnergy efficient, less waste
Unit SizeEA