Fiberlite™ F14-6 x 250LE Fixed Angle Rotor
Fiberlite™ F14-6 x 250LE Fixed Angle Rotor
Actual product may vary
Fiberlite™ F14-6 x 250LE Fixed Angle Rotor
Fiberlite™ F14-6 x 250LE Fixed Angle Rotor
Fiberlite™ F14-6 x 250LE Fixed Angle Rotor
Maximize performance with versatility, speed and a robust, corrosion-free carbon fiber design that supports a range of capacities and applications with the fixed angle rotor.
Have Questions?
Catalog NumberMax. SpeedMax. RCFFor Use With (Equipment)
7500366210,000 rpm (11,000 rpm 230V)15,316 x g (18,533 x g with 230 V)Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Catalog number 75003662
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Max. Speed:
10,000 rpm (11,000 rpm 230V)
Max. RCF:
15,316 x g (18,533 x g with 230 V)
For Use With (Equipment):
Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Increase performance with versatility, speed and a robust, corrosion-free carbon fiber design by using the Thermo Scientific™ Fiberlite™ F14-6 x 250LE Fixed Angle Rotor, which supports a range of capacities and applications.
Fiberlite Rotors:
  • Reduce run times with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design that ensures structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Ordering Information:
Tube adapters are sold separately. Order O-ring Refresher Kit (021-062-153) for replacements. Contact your sales representative for information.
Compatible With:
Thermo Scientific™ Sorvall™ Super T21, Sorvall Legend™ XT, T+, Multifuge™ X3, 3+ Series, Beckman™ GS-6, Allegra™ 6, GP Series.
Recommended for:
Pelleting bacteria or yeast cells and for purifying nucleic acids and cells.
Note:
†Performance may vary by centrifuge model.
Specifications
DescriptionFiberlite F14-6 x 250LE Fixed Angle Rotor for use with Thermo Scientific Sorvall Legend XT, Multifuge X3 Series
TypeFixed Angle Rotor
Capacity6 x 250 mL
Max. RCF15,316 x g (18,533 x g with 230 V)
Max. Speed10,000 rpm (11,000 rpm 230V)
Radius (Metric) Min.3.7 cm
K Factor2737
Max. Tube Length125 mm
Max. Tube Diameter62 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle23°
BiocontainmentYes
Green FeaturesEnergy efficient, less waste
Unit SizeEA