TX-400 4 x 400 mL Swinging Bucket Rotor
TX-400 4 x 400 mL Swinging Bucket Rotor
Actual product may vary
TX-400 4 x 400 mL Swinging Bucket Rotor
Support a wide range of sample sizes and applications with the swinging bucket rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75003656Bucket Lid
75003181Swinging Bucket Rotor
75003655Bucket
Catalog number 75003656
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Bucket Lid
Support a wide range of sample sizes and applications with the Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor.
Ordering Information:
Buckets and lids are sold separately; order rotor body (75003181) and components to complete assembly. For replacement O-rings for lids (set of four), order 75003657. Tube adapters are also available separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Heraeus™ Multifuge™ X1, Heraeus Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Note:
Rotor 75003181 is the replacement for rotor 75003629
Specifications
DescriptionClickSeal Biocontainment Lids
TypeBucket Lid
For Use With (Equipment)Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes
QuantitySet of 4
Unit SizeEA