TX-750 Round Bucket Adapters
TX-750 Round Bucket Adapters
Actual product may vary
TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
75003639
also known as SC75003639
15 mL Conical Tube
75003638
also known as SC75003638
50 mL Conical Tube
75003713100 mL Round-bottom Open-top Tube
7500371984 x 15 mL Blood Collection Tube
750037235/7 mL Blood Collection Tube
750037331.5/2 mL Conical Microtube
75003792250 mL Corning Conical Bottle
Catalog number 75003639
also known as SC75003639
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
15 mL Conical Tube
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
TypeRound Bucket Adapter
Tube TypeConical
Capacity15 mL Conical Tube
For Use With (Equipment)Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube
Tube Size15 mL
Unit SizeEA