M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Actual product may vary
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
750036242 x 3 Standard or 2 x 1 Deepwell Plates
Catalog number 75003624
Price (MYR)
-
Capacity:
2 x 3 Standard or 2 x 1 Deepwell Plates
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Ordering Information:
Order 75003625 for Sealed Carriers, with trays and lids (set of 2), and 75002011 for Replacement Sealing Lids (set of 2). For Replacement O-rings for lids (set of 4 with grease) order 75002012. Contact your Sales Representative for information.
Includes:
Two buckets, two adapters
Compatible with:
Thermo Scientific Sorvall™ 16/16R, Sorvall Lengend™ X1/X1R, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Specifications
DescriptionM-20 Microplate Rotor
TypeSwinging Bucket Rotor
Capacity2 x 3 Standard or 2 x 1 Deepwell Plates
Max. RCF2,272 x g
Max. Speed4,000 rpm
Radius (Metric)12.7 cm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge 16 Centrifuge Series
Angle90°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA