HIGHConic II Aluminum Fixed-Angle Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
HIGHConic II Aluminum Fixed-Angle Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Actual product may vary
HIGHConic II Aluminum Fixed-Angle Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Deliver biocontainment at 10,350rpm with the Thermo Scientific™ HIGHConic™ II Aluminum Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
750036206 x 94 mL
Catalog number 75003620
Price (MYR)
-
Capacity:
6 x 94 mL
Deliver biocontainment at 10,350rpm with the Thermo Scientific™ HIGHConic™ II Aluminum Fixed-Angle Rotor.
Ordering Information:
Adapters are sold separately. Replacement sealing O-rings are also available (in sets of two with grease). Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST 40/40R, Sorvall Legend™ XT/XTR, Megafuge™ 40/40R, Multifuge™ X3/X3R Centrifuge Series.
Specifications
DescriptionHighconic II Fixed-Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity6 x 94 mL
Max. RCF15,090 x g
Max. Speed10,350 rpm
Radius (Metric)12.6 cm
Radius (Metric) Min.6.1 cm
K Factor1713
Max. Tube Length105 mm
Max. Tube Diameter38 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge 40/40R, Multifuge X3/X3R Centrifuge Series.
Angle45°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA