HIGHPlate 6000 Microplate Rotor for 4 L General Purpose X Pro Centrifuges
HIGHPlate 6000 Microplate Rotor for 4 L General Purpose X Pro Centrifuges
Actual product may vary
HIGHPlate 6000 Microplate Rotor for 4 L General Purpose X Pro Centrifuges
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor.
Have Questions?
Catalog NumberCapacity
750036062 x 5 Standard Plates
Catalog number 75003606
Price (MYR)
-
Capacity:
2 x 5 Standard Plates
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor, for use with DNA purification protocols.
Compatible with:
Thermo Scientific™ Sorvall™ Legend™ XT/XTR, Multifuge™ X3/X3R
Specifications
DescriptionHIGHPlate 6000 Microplate Rotor
TypeSwinging Bucket Rotor
Capacity2 x 5 Standard Plates
Max. RCF6,168 x g
Max. Speed6,300 rpm
Radius (Metric)139 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle90°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA