Hematocrit Rotor for Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Hematocrit Rotor for Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Actual product may vary
Hematocrit Rotor for Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Process capillary tubes via centrifugation with the Thermo Scientific™ Hematocrit Rotor.
Have Questions?
Catalog NumberCapacity
7500347324 Capillary Tubes
Catalog number 75003473
Price (MYR)
-
Capacity:
24 Capillary Tubes
Process capillary tubes via centrifugation with the Thermo Scientific™ Hematocrit Rotor.
Features:
  • Holds 24 hematocrit tubes
  • Max. Speed/RCF: 14,800rpm/16,800 x g
Includes: Manual count reader card
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionHematocrit Rotor
MaterialAluminum
TypeRotor
Capacity24 Capillary Tubes
Max. RCF16,800 x g
Max. Speed14,800 rpm
Radius (Metric)8.5 cm
Radius (Metric) Min.2.0 cm
For Use With (Equipment)Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Angle90°
BiocontainmentNo
Unit SizeEA