BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
Actual product may vary
BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
BIOShield™ 1000A Swinging-Bucket Rotor
Safely spin frequently-used tubes up to 7164 x g under sealed conditions using Thermo Scientific™ BIOShield™ 1000A Swinging-Bucket Rotor.
Have Questions?
Catalog NumberDescription
75003182BIOShield 1000A Swinging Bucket Rotor w/Slide Coat anti-friction finish
Catalog number 75003182
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
BIOShield 1000A Swinging Bucket Rotor w/Slide Coat anti-friction finish
Safely spin frequently-used tubes up to 7164 x g under sealed conditions using Thermo Scientific™ BIOShield™ 1000A Swinging-Bucket Rotor.
Includes:
Rotor body, windshield, buckets, sealing lid and rotor manual
Compatible with:
Thermo Scientific™ Sorvall™ ST40/40R, SL40/40R, Sorvall Legend™ XT/XTR, Megafuge™ 40/40R, Multifuge™ X3/X3R Centrifuge Series
Recommended for:
Blood tube separations, cell culture and microbiological applications
Ordering Information:
Tube adapters, sold in sets of four, are sold separately. Contact your sales representative for more information.
Specifications
DescriptionBIOShield 1000A Swinging Bucket Rotor w/Slide Coat anti-friction finish
TypeSwinging Bucket Rotor
Capacity4 x 250 mL
Max. RCF7,164 x g
Max. Speed6,000 rpm
Radius (Metric)178 mm
Max. Tube Length130 mm
Max. Tube Diameter63 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40 and SL 40/40F Centrifuges
Angle90°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA