Sorvall™ Stratos Microplate Rotor
Sorvall™ Stratos Microplate Rotor
Actual product may vary
Sorvall™ Stratos Microplate Rotor
2 x 2 microplate capacity
Have Questions?
Catalog NumberCapacity
750030482 x 1 mL
Catalog number 75003048
HSN 84219100
Price (MYR)
0.00
EA
Add to cart
Capacity:
2 x 1 mL
Price (MYR)
0.00
EA
Add to cart
For use with Thermo Scientific Sorvall Stratos Centrifuges.
Specifications
TypeMicrotiter Plate Rotor
Capacity2 x 1 mL
Max. RCF1,370 x g
Max. Speed3,000 rpm
Radius (Metric)1.36 cm
For Use With (Equipment)Thermo Scientific™ Sorvall™ Stratus Centrifuge
Certifications/ComplianceBiocontainment approved
Unit SizeEA