4 x 180mL Swinging Bucket Rotor
4 x 180mL Swinging Bucket Rotor
Actual product may vary
4 x 180mL Swinging Bucket Rotor
Incorporate the Thermo Scientific™ 4 x 180mL Swinging Bucket Rotor with Sorvall™ Stratos™ rotors and adapters.
Have Questions?
Catalog NumberCapacity
75003047
also known as 75003047 
4 x 180 mL
Catalog number 75003047
also known as 75003047 
Price (MYR)
-
Capacity:
4 x 180 mL
Incorporate the Thermo Scientific™ 4 x 180mL Swinging Bucket Rotor with Sorvall™ Stratos™ rotors and adapters.
  • Capacity: 4 × 180mL (0.72L)
  • For 120V: Maximum Speed/RCF: 4500rpm/3940 x g
  • For 208V: Maximum Speed/RCF: 5000rpm/4863 x g
Specifications
Description4 x 180mL Swinging Bucket Rotor
TypeSwinging Bucket Rotor
Capacity4 x 180 mL
Max. RCF3,940 x g (4,863 x g with 208 V)
Max. Speed4,500 rpm (5,000 rpm 208 V)
Radius (Metric)17.4 mm
For Use With (Equipment)Thermo Scientific Sorvall Stratos and Stratos Biofuge Centrifuges
Unit SizeEA