T29-8 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
T29-8 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
T29-8 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Perform applications with a new level of high g-force (up to 100,605 x g) using this superspeed, fixed-angle rotor.
Have Questions?
Catalog NumberCapacity
750030098 x 50 mL
Catalog number 75003009
Price (MYR)
108,757.00
EA
Add to cart
Capacity:
8 x 50 mL
Price (MYR)
108,757.00
EA
Add to cart
Carry out applications with a new level of high g-force (up to 100,605 x g) in superspeed using the Thermo Scientific™ T29-8 x 50mL Fixed-Angle Rotor. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange, to Thermo Scientific™ Auto-ID™ instant rotor identification, to speed handles on each rotor lid—shorten run set-up time while providing peace-of-mind that the rotor is secure.
Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionT29-8x50 Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity8 x 50 mL
Max. RCF100,605 x g
Max. Speed29,000 rpm
Radius (Metric)107 mm
Radius (Metric) Min.33 mm
K Factor354
Max. Tube Length108 mm
Max. Tube Diameter29 mm
Net Weight18.5 lb. (8.4 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle34°
BiocontainmentYes
Unit SizeEA