A27-6 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
A27-6 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
A27-6 x 50 Fixed Angle Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Maintain low sample temperatures at all speeds while experiencing high RCF performance with the Thermo Scientific™ A27-6 x 50mL Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
750030076 x 50 mL
Catalog number 75003007
Price (MYR)
48,147.00
EA
Add to cart
Capacity:
6 x 50 mL
Price (MYR)
48,147.00
EA
Add to cart
Maintain low sample temperatures at all speeds while experiencing high RCF performance with the Thermo Scientific™ A27-6 x 50mL Fixed-Angle Rotor, which features rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange, to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid— to shorten run set-up time while providing peace of mind that the rotor is secure.
Auto-Lock Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionA27-6x50 Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity6 x 50 mL
Max. RCF79,057 x g
Max. Speed27,000 rpm
Radius (Metric)97 mm
Radius (Metric) Min.24 mm
K Factor485
Max. Tube Length108 mm
Max. Tube Diameter29 mm
Net Weight12.3 lb. (5.58 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle34°
BiocontainmentYes
Unit SizeEA