A23-6 x 100 Fixed Angle Rotor
A23-6 x 100 Fixed Angle Rotor
Actual product may vary
Thermo Scientific
A23-6 x 100 Fixed Angle Rotor
Perform a variety of applications due to multiple adapters for smaller tubes with the Thermo Scientific™ A23-6x100 Fixed Angle Rotor.
Have Questions?
Catalog NumberCapacity
750030066 x 100 mL
Catalog number 75003006
Price (MYR)
40,595.00
EA
Add to cart
Capacity:
6 x 100 mL
Price (MYR)
40,595.00
EA
Add to cart
Perform a variety of applications due to multiple adapters for smaller tubes with the Thermo Scientific™ A23-6x100 Fixed Angle Rotor. Innovations — Thermo Scentific™ Auto-Lock™ rotor exchange, Thermo Scentific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid — shorten run set-up time while providing peace-of-mind that the rotor is secure.
Auto-Lock Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionA23-6x100 Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity6 x 100 mL
Max. RCF62,976 x g
Max. Speed23,500 rpm
Radius (Metric)102 mm
Radius (Metric) Min.34 mm
K Factor503
Max. Tube Length105 mm
Max. Tube Diameter38 mm
Net Weight15.8 lb. (7.2 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle25°
BiocontainmentYes
Unit SizeEA