Specifications
DescriptionA22-24x16 Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity24 x 16 mL
Max. RCF60,063 x g (Outer), 60,063 x g (Inner)
Max. Speed22,000 rpm
Radius (Metric)111 mm Outer, 111 mm Inner
Radius (Metric) Min.54 mm Outer, 39 mm Inner
K Factor377 (Outer); 547 (Inner)
Max. Tube Length102 mm
Max. Tube Diameter18 mm
Net Weight17.9 lb. (7.7 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle28/40°
BiocontainmentYes
Unit SizeEA