A21-24 x 15c Fixed Angle Rotor
A21-24 x 15c Fixed Angle Rotor
Actual product may vary
A21-24 x 15c Fixed Angle Rotor
Carry out subcellular separations and fractionations with 15mL conical tubes using the Thermo Scientific™ A21-24 x 15c Fixed Angle Rotor.
Have Questions?
Catalog NumberCapacity
7500300424 x 15 mL
Catalog number 75003004
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
24 x 15 mL
Carry out subcellular separations and fractionations with 15mL conical tubes using the Thermo Scientific™ A21-24 x 15c Fixed Angle Rotor. Innovations —from Thermo Scientific™ Auto-Lock™ rotor exchange, to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid — shorten run set-up time while providing peace-of-mind that the rotor is secure.
Auto-Lock Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionA21-24x15c Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity24 x 15 mL
Max. RCF63,049 x g (Outer), 63,049 x g (Inner)
Max. Speed21,500 rpm
Radius (Metric)158 mm
Radius (Metric) Min.57 mm
K Factor379
Max. Tube Length121.5 mm
Max. Tube Diameter16.5 mm
Net Weight19.1 lb. (8.7 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle42/30°
BiocontainmentYes
Unit SizeEA