BIOFlex HS Swinging-Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
BIOFlex HS Swinging-Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
BIOFlex HS Swinging-Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Support more than 30 tube types, from microtubes to conical tubes to 400mL bottles, with the versatile adapter options of the BIOFlex HS Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
750030024 x 400 mL
Catalog number 75003002
Price (MYR)
66,442.00
EA
Add to cart
Capacity:
4 x 400 mL
Price (MYR)
66,442.00
EA
Add to cart
Support more than 30 tube types, including microtubes, conical tubes and 400mL bottles, with the versatile adapter options of the Thermo Scientific™ BIOFlex™ HS Swinging-Bucket Rotor. Rotor innovation—from Thermo Scientific™ Auto-Lock™ rotor exchange, to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid—shorten run set-up time while providing peace of mind that the rotor is secure.
Auto-Lock Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionBIOFlex HS Swinging Bucket Rotor
MaterialStainless Steel Rotor Body, Aluminum Bucket
TypeSwinging Bucket Rotor
Capacity4 x 400 mL
Max. RCF10,025 x g
Max. Speed7,000 rpm
Radius (Metric)183 mm
Radius (Metric) Min.71 mm
K Factor4,889
Max. Tube Length125 mm
Max. Tube Diameter80 mm
Net Weight9.6 lb. (4.36 kg) Rotor Cross, 16.8 lb. (7.64 kg) Cross with Buckets
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
AngleHorizontal
BiocontainmentYes
Unit SizeEA