TZ-32 Zonal Rotor
TZ-32 Zonal Rotor
Actual product may vary
TZ-32 Zonal Rotor
TZ-32 Zonal Rotor
TZ-32 Zonal Rotor
Thermo Scientific™ TZ-32 Zonal Rotor, delivering maximum flexibility at 32,000rpm and 102,000 xg, is designed with efficiency for large volume separations using density gradient.
Have Questions?
Catalog number 75000008
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Thermo Scientific™ TZ-32 Zonal Rotor, delivering maximum flexibility at 32,000rpm and 102,000 xg, is designed with efficiency for large volume separations using density gradient.
Thermo Scientific™ TZ-32 Zonal Rotor is ideal for the development and pilot scale manufacturing of vaccines, gene therapies and other bio-products. Its batch volume is 1,690 mL. Working on the Sorvall WX+ series ultraspeed centrifuge, it offers cost-effective, reliable and proven solution for isolation, purification and concentration of viruses, viral vectors and proteins.
Specifications
MaterialTitanium Alloy
TypeZonal Rotor
Capacity1,690 mL
Max. RCF102,000 xg
Max. Speed32,000 rpm
Net Weight15.4 kg
For Use With (Equipment)Thermo Scientific Sorvall WX+ series ultraspeed centrifuges
Unit SizeEA