ProPac™ SCX-20 Strong Cation Exchange HPLC Column
Product Image
ProPac™ SCX-20 Strong Cation Exchange HPLC Column
Meet today's demands in protein separations using this high resolution strong cation exchange (SCX) HPLC column.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
74628250 mm
7464350 mm
Catalog number 74628
Price (MYR)
-
Length (Metric):
250 mm
Meet today's demands for high resolution protein separations using the Thermo Scientific™ ProPac™ SCX-20 column. This strong cation exchange (SCX) column is designed specifically for fast mass transfer and high efficiency, for a combination that results high resolution separations.
The stationary phase is composed of a 10μm, non-porous, solvent compatible ethylvinylbenzene-divinylbenzene copolymer beads that are uniformly coated with a highly hydrophilic layer to reduce nonspecific interactions between the surface and the biopolymer. The surface of the beads is grafted with SCX functionalized polymer layer providing well-controlled graft density and uniform distribution of chain lengths.
High resolution separation of proteins
  • Grafted cation-exchange surface provides pH-based selectivity control
  • Fast mass transfer for high-efficiency separation
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)250 mm
Max. Pressure3000 psi (207 bar)
Particle Size10 μm
pH2 to 12
QuantityEach
Temperature60°C
Stationary PhaseSulfonate
Product LineProPac
Packing MaterialEthylvinylbenzene Divinylbenzene
Column TypeIon Exchange
PhaseIon Exchange
TypeAnalytical Column
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEach