Accucore™ XL C8 HPLC Columns
Accucore™ XL C8 HPLC Columns
Actual product may vary
Accucore™ XL C8 HPLC Columns
Improve laboratory productivity using Thermo Scientific™ Accucore™ XL C8 Columns.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
74204-154630150 mm4.6 mm
Catalog number 74204-154630
Price (MYR)
-
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Optimize conventional HPLC methods with Thermo Scientific™ Accucore™ XL C8 columns. Rugged 4μm solid-core particles ensure very high efficiencies within standard HPLC pressure limits. The high coverage C8 bonded phase provides optimal retention of analytes with moderate hydrophobicity. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Lower Hydrophobic Retention
  • Recommended for moderately hydrophobic analytes or when a less hydrophobic phase provides optimum retention
High-Efficiency Separations for Enhanced Resolution and Sensitivity
  • Core-enhanced technology 4μm particles and narrow particle size distribution reduce band broadening and improve separation efficiencies for conventional HPLC methods.
  • High surface coverage of silica minimizes secondary interactions and peak tailing.
Same System, Same Method, Better Results
  • Solid-core 4μm particle design means that no changes to the method or system are required.
Excellent Reproducibility and Robustness
  • Advanced bonding technology
  • Automated packing process
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1.5 mL/min
For Use WithHPLC
Length (Metric)150 mm
Max. Pressure8700 psi (600 bar)
Particle Size4 μm
pH2 to 9
Pore Size80 Å
Surface Area130 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load4%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach