BioBasic™ AX HPLC Columns
BioBasic™ AX HPLC Columns
Actual product may vary
BioBasic™ AX HPLC Columns
Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
73105-154630150 mm4.6 mm
Catalog number 73105-154630
Price (MYR)
-
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.
  • 5μm particle size, 300Å pore size
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • Capacity of 0.22mEq/g; 3% carbon load
  • Use for normal phase and hydrophilic interaction chromatography (HILIC)
  • Stable under demanding pH conditions (2 to 8 pH range)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
HydrophobicityLow
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeN/A
Stationary PhasePolyethyleneimine
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load3%
Column TypeMixed Mode
EndcappedNo
PhaseNormal Phase, HILIC, Ion Exchange
TechniqueBio-LC
Unit SizeEach