Check Order Status
16 in. Single Blade Ultra Hygiene Bench Squeegee
Actual product may vary
16 in. Single Blade Ultra Hygiene Bench Squeegee
Vikan™ 16 in. Single Blade Ultra Hygiene Bench Squeegee is ideal for removing water or food debris from food contact surfaces.
Have Questions?
Catalog Number
Color
71401
Pink
Catalog number
71401
Price (MYR)
-
Color:
Pink
One-piece construction
Single-blade squeegee
Ultra-hygienic design that minimizes areas for water and food soils to collect
Effective in areas requiring the highest level of hygiene
The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces
Hygiene squeegees are a good foundation for any HACCP or food safety program
A splash guard helps prevent water from reaching the dried surface
Color-coded
Specifications
Certifications/Compliance
FDA Compliant
Color
Pink
Description
Thread type Euro Hygienic
Material
Polypropylene, Rubber
Quantity
1 Ea.
Size
Large
Width (English)
3.5 in.