Oxidized-CaM Kinase II (Met281/282) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Oxidized-CaM Kinase II (Met281/282) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Actual product may vary
Oxidized-CaM Kinase II (Met281/282) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Have Questions?
Catalog number 71387
Price (MYR)
-
This Anti-oxidized-CaM Kinase II (Met281/282) Antibody is validated for use in western blot, IC for the detection of oxidized-CaM Kinase II (Met281/282).
For Research Use Only
Specifications
Antigenoxidized-CaM Kinase II (Met281/282)
ApplicationsImmunocytochemistry, Western Blot
ClassificationPolyclonal
ConjugateUnconjugated
FormulationUnpurified rabbit serum containing 0.05% NaN3.
Gene SymbolsCAMKK; CAMKKB; EC 2.7.11.17 ; KIAA0787; MGC15254
Host SpeciesRabbit
ImmunogenSynthetic peptide encompassing and including oxidized Met281/282.
Quantity100 μL
Primary or SecondaryPrimary
Gene ID (Entrez)NP_742127
Target SpeciesHuman, Mouse, Rat
FormSerum
Unit SizeEach
Contents & Storage
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles