Nalgene™ Utility Carriers
Nalgene™ Utility Carriers
Actual product may vary
Nalgene™ Utility Carriers
Carry glassware, samples or tools with this utility carrier, featuring resilient construction with molded-in center divider and handle.
Have Questions?
Catalog NumberMaterial
7130-0010Polyethylene
Catalog number 7130-0010
Price (MYR)
1,113.00
CS
Add to cart
Material:
Polyethylene
Price (MYR)
1,113.00
CS
Add to cart
Carry glassware, samples or tools with Thermo Scientific™ Nalgene™ Utility Carriers, featuring resilient construction with molded-in center divider and handle.
  • Choice of polyethylene or autoclavable polypropylene
  • Exterior dimensions (LxWxH): 14.875 x 9.5 x 4.5 in. (38 x 24 x 11cm)
Specifications
Product TypeBottle Carrier
Certifications/ComplianceOSHA Standard 29CFR1910
DescriptionUtility Carrier
AutoclavableAutoclavable
MaterialPolyethylene
For Use With (Application)For carrying glassware, samples, tools
No. per Case6
No. per Pack1
Dimensions (L x W x H)14.875 x 9.5 x 4.5 in. (38 x 24 x 11cm)
Length (Metric)38 cm
Length (English)14.875 in.
Width (Metric)24 cm
Width (English)9.5 in.
Height (Metric)11 cm
Height (English)4.5 in.
Unit SizeCS