BetaSil™ C18 HPLC Columns, 5μm Particle Size
BetaSil™ C18 HPLC Columns, 5μm Particle Size
Actual product may vary
BetaSil™ C18 HPLC Columns, 5μm Particle Size
High surface area with high bonded phase coverage
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
70105-254630250 mm4.6 mm
Catalog number 70105-254630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
High surface area with high bonded phase coverage
  • Thermo Scientific chemistry-bonded on 100&Angst; silica
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Film Thickness5 μm
For Use WithLC/MS Separation
Functionality/Interaction TypeStandard
Length (Metric)250 mm
Particle Size5 μm
pH2 to 9
Pore Size100 Å
Surface Area310 m²/g
Surface CoveragePhase coverage
USP TypeL1
Stationary PhaseC18
Product LineBeta
Packing MaterialSilica
Carbon Load20%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeColumn
Unit SizeEach