Hydroguard MXT Guard Columns and Transfer Lines
Hydroguard MXT Guard Columns and Transfer Lines
Actual product may vary
Hydroguard MXT Guard Columns and Transfer Lines
Hydroguard MXT Guard Columns and Transfer Lines
Hydroguard MXT Guard Columns and Transfer Lines
Extends analytical column lifetime by preventing degradation from harsh steam-cleaning water injections.
Have Questions?
Catalog NumberLength (Metric)
70087150 mm, 150 mm
Catalog number 70087
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm, 150 mm
  • Passivated stainless steel
  • Maximum temperature: 325°C
Specifications
TypeGuard Column Transfer Line
Diameter (Metric) Inner0.53 mm
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 to 0.5 mL/min
Length (Metric)150 mm, 150 mm
Max. Pressure3000 psi (207 bar) aqueous, 2000 psi (140 bar) organic
Metal Impurity<10.0 ppm Total Metal, <3.0 ppm Individual Metal
Particle ShapeSpherical
Particle Size3 μm
Particle Size Distribution<1.3
pH2 to 8
Pore Size120 Å
Pore Volume1 mL/g
QuantityEach
Surface Area300 m²/g
Surface Coverage2.7 mmol/m²
Temperature60°C
Stationary PhaseProprietary
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load17%
Column TypeSpecialty/Application Specific
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Green FeaturesResponsibly packaged
Unit SizeEach