Acclaim™ Organic Acid HPLC Columns
Acclaim™ Organic Acid HPLC Columns
Actual product may vary
Acclaim™ Organic Acid HPLC Columns
Use for separation of hydrophilic, aliphatic, and aromatic organic acids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
70086150 mm3 mm3 μm
062902
also known as CF1/4CT52R394J
250 mm4 mm5 μm
062903150 mm4 mm5 μm
Catalog number 70086
Price (MYR)
-
Length (Metric):
150 mm
Diameter (Metric):
3 mm
Particle Size:
3 μm
For unparalleled performance in separating hydrophilic, aliphatic, and aromatic organic acids use Thermo Scientific™ Acclaim™ Organic Acid LC Columns. These high-efficiency, reversed-phase silica columns are compatible with 100% aqueous mobile phases and have excellent hydrolytic stability at low pHs for excellent selectivity in the separation of organic acids. Achieve high-resolution separations with fast run times and minimal sample preparation.
High-Efficiency, High-Throughput Organic Acid Analysis
  • Ideal selectivity for small hydrophilic organic acids, C1 to C7 aliphatic acids and hydrophilic aromatic acids
  • Excellent column efficiency and peak shapes for organic acids
  • Hydrolytic stability at low-pH conditions for optimum reversed-phase retention of organic acids
  • Use-tested to guarantee consistent hydrophilic organic acid separations
  • Packed in metal-free PEEK column bodies to eliminate unwanted interaction between the analytes and the column body
  • Available in 3.0 and 5.0μm particle sizes and 2.1, 3.0, and 4.0mm diameters, with 150 and 250mm lengths
Recommended for analysis and quality testing of foods, beverages, pharmaceuticals, chemical intermediates and environmental samples.
About the Family of Acclaim Specialty HPLC Columns
Acclaim specialty columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease-of-use.
  • Use-tested to guarantee consistent separations
  • Ideal selectivity for separating specific target analytes
  • Excellent peak shapes
  • Performance tested to ensure high quality and reliability for specific chromatographic separations
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.3 to 0.8 mL/min
Length (Metric)150 mm
Max. Pressure3000 psi (207 bar) aqueous, 2000 psi (140 bar) organic
Metal Impurity<10.0 ppm Total Metal, <3.0 ppm Individual Metal
Particle ShapeSpherical
Particle Size3 μm
Particle Size Distribution<1.3
pH2 to 8
Pore Size120 Å
Pore Volume1 mL/g
QuantityEach
Surface Area300 m²/g
Surface Coverage2.7 mmol/m²
Temperature60°C
Stationary PhaseProprietary
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load17%
Column TypeSpecialty/Application Specific
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Green FeaturesResponsibly packaged
Unit SizeEach