Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
69700 | 50 Cups, 50 Cups |
Thermo Scientific™ Pierce Microcentrifuge Spin Cups and Spin Columns are convenient tools for manipulating small volumes of affinity supports for protein purification.
These microcentrifuge spin cups and spin columns are convenient tools for manipulating small volumes of affinity supports (5 to 500μL) for immunoprecipitation (IP) and pull-down purification assays. Five styles of polypropylene columns and matching collection tubes offer a variety of options to suit particular types of experimental needs. Add the affinity resin and sample to one of the columns and use a microcentrifuge to efficiently wash away contaminants and elute your purified sample without losing any resin in the process. Spin columns allow you to affinity-purify more protein in less time.
Highlights:
Spin Cups-Paper Filter with Collection Tubes (Part No. 69700):
Spin Cups-Cellulose Acetate Filter with Collection Tubes (Part No. 69702):
Spin Columns-Screw Cap (Part No. 69705):
Spin Columns-Snap Cap with Collection Tubes (Part No. 69725):
Micro-Spin Columns (Part No. 89879):
Microcentrifuge Tubes (Part No. 69715):
Microcentrifuge Tubes (Part No. 69720):