KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
Actual product may vary
KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
KGA20 Hooded Coveralls with Boots
Coveralls with attached hood and boots provide lightweight protection against heavy dust and limited splashes.
Have Questions?
Catalog NumberSize
689814X-Large
Catalog number 68981
Price (MYR)
-
Size:
4X-Large
  • For Non-Hazardous Particulate Protection
  • Comfortable, lightweight, SMS fabric is breathable and allows moisture to escape
  • Hood and Boot, Elastic Wrist and Ankle Design
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
Specifications
Product TypeGeneral Purpose Coveralls
Size4X-Large
GenderUnisex
MaterialSMS Fabric
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Closure TypeFront Zipper
SterilityNon-sterile
ResistanceDust, Liquid
Certifications/ComplianceANSI/ISEA 101-1996
Certifications/Compliance (Europe)CE Category I PPE
Latex FreeYes
ModelKleenGuard™ KGA20
For Use With (Application)Agriculture, Food Processing, General Maintenances, Laboratories
Unit SizeEach