KGA20 Hooded Coveralls
KGA20 Hooded Coveralls
Actual product may vary
KGA20 Hooded Coveralls
KGA20 Hooded Coveralls
KGA20 Hooded Coveralls
KGA20 Hooded Coveralls
KGA20 Hooded Coveralls
Coveralls with attached hood provide lightweight protection against heavy dust and limited splashes.
Have Questions?
Catalog NumberSize
68973X-Large
Catalog number 68973
Price (MYR)
-
Size:
X-Large
  • Comfortable, lightweight, SMS fabric is breathable and allows moisture to escape
  • Hooded, Elastic Wrist and Ankle Design
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
Specifications
Product TypeGeneral Purpose Coveralls
SizeX-Large
GenderUnisex
MaterialSMS Fabric
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Closure TypeFront Zipper
Seam TypeStitched
SterilityNon-sterile
ResistanceDust, Liquid
Certifications/ComplianceANSI/ISEA 101-1996
Certifications/Compliance (Europe)CE Category I PPE
Latex FreeYes
ModelKleenGuard™ KGA20
For Use With (Application)Agriculture, Food Processing, General Maintenances, Laboratories
Unit SizeEach