GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
Actual product may vary
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
Measure and monitor the flow of all types of gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter.
Have Questions?
Catalog NumberAccuracy
66002-010±2% of Flow or ± 0.05 mL/min., whichever is greater
Catalog number 66002-010
Price (MYR)
-
Accuracy:
±2% of Flow or ± 0.05 mL/min., whichever is greater
Measure and monitor the flow of all types of laboratoty gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter. Specifically designed for use with GC systems, this CE-certified flowmeter has a measurement range of 0.5–500mL/min and measures the volumetric flow of any gas with an accuracy of +/- 2% or +/- 0.2mL/min, whichever is greater. Its easy-to-use interface features an over-range warning indicator and automatic shutoff, and its explosion-proof rating allows use with flammable and explosive gases. The compact ergonomic design enables use as a hand-held or benchtop unit and features side grips for added durability.
Additional Product Information
  • Data output via USB port
  • Calibration: traceable to NIST primary standards
  • Uses two AA batteries
  • Recalibration service available
Specifications
Accuracy±2% of Flow or ± 0.05 mL/min., whichever is greater
Product TypeNon-magnetic Cell Isolation Kit
DescriptionElectronic Gas Flowmeter
Donor SourceHuman Whole Blood
For Use With (Application)Isolation of label-free cells
Sufficient For9 mL Whole Blood
Quantity10 Kits, Human
Unit SizeEach
Contents & Storage
2°C to 8°C