Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Actual product may vary
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
66002-003Small Probe Adapter
66002-002Soft-sided Carry Case
Catalog number 66002-003
Price (MYR)
-
Type:
Small Probe Adapter
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector GLD Pro.
Soft-sided carrying case also for use with GFM Pro Handheld Equipment
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeSmall Probe Adapter
For Use With (Equipment)GLD Pro Gas Leak Detector
Unit SizeEach