Polypropylene Flaoting Microtube Rack
Polypropylene Flaoting Microtube Rack
Actual product may vary
Polypropylene Flaoting Microtube Rack
Float Rack that can be used with variety of tubes.
Have Questions?
Catalog NumberPositions
6320196
Catalog number 63201
Price (MYR)
-
Positions:
96
Floating microtube racks are ideal for thawing, cooling, or incubating samples in water or ice baths.
  • Can be used at a wide range of temperatures from -70°C to 100°C
  • Tubes are supported below the lip while conical ends are immersed in water
  • Racks feature a handle for easy removal from water bath, as well as a molded-in alpha-numeric grid for identification of samples
Specifications
IncludesRack
Product TypeMicrotube Rack
Array4 x 4
AutoclavableNot Autoclavable
ColorWhite
HoldsMicrotubes
MaterialPolypropylene
Positions96
ShapeSquare
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)Microtubes
Length (Metric)21.59 cm
Width (Metric)21.59 cm
Height (Metric)12.7 cm
Unit SizeEach