Check Order Status
Filter Disc, Surfaces Plane, Ground Cylindrical, 290 mm
Filter Disc, Surfaces Plane, Ground Cylindrical, 290 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Have Questions?
Change view
Catalog Number
Porosity
62903
3
62902
2
2 Options
Catalog number
62903
Price (MYR)
-
Porosity:
3
Highly resistant to most chemicals and temperatures up to 500°C
The biplane version may be required in specific installation circumstances
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
Porosity
3
Thickness (Metric)
18.0 mm ± 0.5 mm
Diameter (Metric)
290 mm