Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Actual product may vary
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liner
Find a variety of uses for these polyethylene bench/drawer liners, such as sink liners or titration backgrounds.
Have Questions?
Catalog NumberWidth (English)
6283-125012 in.
Catalog number 6283-1250
Price (MYR)
1,926.00
CS
Add to cart
Width (English):
12 in.
Price (MYR)
1,926.00
CS
Add to cart
Find a variety of uses for Thermo Scientific™ Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liners, such as sink liners or titration backgrounds. These liners can be cut to any size and stapled, tacked or taped. They provide cushioning and liquid barrier protection.
  • Made of 0.125 in. (3.2mm) thick, closed-cell polyethylene foam to cushion benchtop items
  • Smooth and resilient, this non-absorbent pad is versatile in your lab — use for a multitude of purposes
  • Excellent chemical resistance provides protection against spills wherever placed
  • Can be cut to any size to fit benches, shelves, drawers — anywhere protection is needed against spills, vibration, heat, dirt or breakage
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Product TypeBench/Drawer Liner
MaterialPolyethylene
Length (English)50 ft.
Length (Metric)15.2 m
Width (English)12 in.
Width (Metric)31 cm
DescriptionPolyethylene bench/drawer liners
No. per Case2
No. per Pack1
Item DescriptionLiners Clean Sheets XLPE 30.5 cm x 15 m
Unit SizeCS