Temperature Chart Recorders
Temperature Chart Recorders
Actual product may vary
Temperature Chart Recorders
Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.
Have Questions?
Catalog NumberFor Use With (Equipment)Temperature RangeType
6183-7-40°C to + 38°C-40°C to +38°CBuilt-In
Catalog number 6183-7
Price (MYR)
14,472.00
EA
Add to cart
For Use With (Equipment):
-40°C to + 38°C
Temperature Range:
-40°C to +38°C
Type:
Built-In
Price (MYR)
14,472.00
EA
Add to cart
Record seven-day history on 6 in. charts using Thermo Scientific™ Temperature Chart Recorders, for use with most freezers and refrigerators.
Features:
  • Seven-day chart recorders record temperatures from -100° to 0°C (-40° to +38° on 6183-7)
  • 6 in. (15.2cm) chart
  • Use pressure-sensitive paper, require no inking
  • Freestanding model is customer installed and must be installed by a qualified professional
  • Panel-mounted model is factory installed; order when purchasing refrigerator or freezer
Ordering Information:
Available in range of voltages; specify voltage when ordering.
Includes:
Pack of 50 charts
Specifications
DescriptionSeven-day chart recorders
TypeBuilt-In
Temperature Range-40°C to +38°C
StyleBuilt-in
For Use With (Equipment)-40°C to + 38°C
Static Holding Time7 Days
Diameter (English)6 in.
Diameter (Metric)15.2 cm
Monitoring OptionsRecorder
Unit SizeEA