Check Order Status
Isotemp™ Chiller Installation Kit
Isotemp™ Chiller Installation Kit
Installation kit
Have Questions?
Catalog number
611000000108
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
For use with Fisher Scientific Isotemp Recirculating Chillers
Specifications
Type
Installation Kit
Includes
0.5 x 0.5 in. MPT fittings, 25 ft. of 0.62 in. I.D. Polybraid hose, hose clamps and hose insulation