Fluid Filter Kit
Fluid Filter Kit
Actual product may vary
Fluid Filter Kit
Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)Partial Flow
611000000102
also known as 611000000102N
Replacement filter bags (quantity 2)ThermoFlex™ 7500 or 24000100 micron
Catalog number 611000000102
also known as 611000000102N
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Replacement filter bags (quantity 2)
For Use With (Equipment):
ThermoFlex™ 7500 or 24000
Partial Flow:
100 micron
Thermo Scientific™ Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
Specifications
DescriptionReplacement filter bags (quantity 2)
For Use With (Equipment)ThermoFlex™ 7500 or 24000
TypeReplacement Filter Bags
Partial Flow100 micron
Unit SizeEA