Terry Tool Tubing Cutters
Terry Tool Tubing Cutters
Actual product may vary
Terry Tool Tubing Cutters
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Have Questions?
Catalog NumberFor Use With (Equipment)
60182-5091/16 in. Stainless Steel Tubing
Catalog number 60182-509
Price (MYR)
-
For Use With (Equipment):
1/16 in. Stainless Steel Tubing
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeTubing Cutter
DescriptionCuts 0.0625 stainless steel tubing
For Use With (Equipment)1/16 in. Stainless Steel Tubing
Unit SizeEach