Deactivated Glass Wool
Product Image
Deactivated Glass Wool
Pack GC injection port liners with deactivated glass wool for good homogenization, volatilization of high boiling point compounds, and minimal column contamination.
Have Questions?
Catalog NumberDescription
60180-785Deactivated Glass Wool 10 g
Catalog number 60180-785
Price (MYR)
-
Description:
Deactivated Glass Wool 10 g
Pack your GC injection port liner with Thermo Scientific™ deactivated glass wool for good homogenization and volatilization of high boiling point compounds within the liner. The glass wool acts as a barrier to prevent non-volatile sample matrix compounds and septa particles from entering GC columns, minimizing contamination and increasing precision and reproducibility.
Specifications
DescriptionDeactivated Glass Wool 10 g
Quantity10 g
Packing MaterialDeactivated Glass Wool
For Use With (Equipment)GC Liners
Unit SizeEach