PTFE One-Piece HPLC Column Connector
PTFE One-Piece HPLC Column Connector
Actual product may vary
PTFE One-Piece HPLC Column Connector
PTFE One-Piece HPLC Column Connector
PTFE One-Piece HPLC Column Connector
PTFE One-Piece HPLC Column Connector
PTFE One-Piece HPLC Column Connector
Ensure leak-free column connections and optimize liquid chromatography performance with the inert and biocompatible Thermo Scientific™ PTFE One-Piece HPLC Column Connector.
Have Questions?
Catalog number 60170-370
Price (MYR)
-
Minimize HPLC system dead volume and ensure optimal separation efficiencies with the Thermo Scientific™ PTFE One-Piece HPLC Column Connector. Ideal for coupling analytical and guard columns with 10–32 threads, it provides fingertight, leak-free connections. The material is inert and biocompatible for a wide range of applications.
Specifications
TypeCoupler
DescriptionOne-piece column coupler
MaterialPEEK
For Use With (Equipment)Analytical and Guard Columns with 10 to 32 Threads
Unit SizeEach