HyperSep™ Pressure Manifold Installation Kit
HyperSep™ Pressure Manifold Installation Kit
Actual product may vary
HyperSep™ Pressure Manifold Installation Kit
Obtain all the parts needed to set up your Thermo Scientific™ HyperSep™ Positive Pressure Manifold.
Have Questions?
Catalog NumberDescription
60104-272Installation Kit, 1 pk
Catalog number 60104-272
Price (MYR)
-
Description:
Installation Kit, 1 pk
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Installation Kit for all the parts needed to set up your manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
Specifications
DescriptionInstallation Kit, 1 pk
For Use With (Application)SPE
For Use With (Equipment)Positive Pressure Manifold
Product LineHyperSep
TypeInstallation Kit
Unit SizeEach