Vial Rack
Vial Rack
Actual product may vary
Thermo Scientific
Vial Rack
Safely store your vials in this autoclavable rack.
Have Questions?
Catalog NumberFor Use With (Equipment)Diameter (Metric) Inner
6.51018 mL Vials17 mm
Catalog number 6.5101
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
8 mL Vials
Diameter (Metric) Inner:
17 mm
  • Sturdy, economical and convenient racks for samples stored in vials.
  • Autoclavable at 120°C for 20 minutes.
  • Can be stacked when not filled, for easy storage.
  • Each well features a drainage hole at the bottom.
  • Available in two sizes.
Warning: Cancer and reproductive harm
Specifications
Includes5 Racks
Product TypeVial Rack
Array15 x 6
AutoclavableAutoclavable
ColorBlue
HoldsVials
MaterialPolypropylene
Positions90
ShapeRectangular
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)8 mL Vials
SizeSmall
Width (Metric)145 mm
Height (Metric)30 mm
Diameter (Metric) Inner17 mm
Unit SizeEach