Deionized Water Support for Recirculating Chillers
Deionized Water Support for Recirculating Chillers
Actual product may vary
Deionized Water Support for Recirculating Chillers
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Support for Recirculating Chillers.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)Includes
598000000010DI cartridgeThermoFlex™ Chiller equipped with IDI (not intended for units with DIC)DI cartridge for all ThermoFlex equipped with a Basic controller and Internal DI (IDI). Not intended for units with DIC
Catalog number 598000000010
Price (MYR)
-
Description:
DI cartridge
For Use With (Equipment):
ThermoFlex™ Chiller equipped with IDI (not intended for units with DIC)
Includes:
DI cartridge for all ThermoFlex equipped with a Basic controller and Internal DI (IDI). Not intended for units with DIC
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Packages.
  • Maintains a water resistivity level between 1 and 3 MOhmcm
Includes: Kits include a filter canister, DI media, plumbing, connectors, filter, and a LED indication to let you know when the media needs to be replaced. DI Cartridges include only the replacement DI media.
Specifications
DescriptionDI cartridge
For Use With (Equipment)ThermoFlex™ Chiller equipped with IDI (not intended for units with DIC)
TypeDeionized Water Support
IncludesDI cartridge for all ThermoFlex equipped with a Basic controller and Internal DI (IDI). Not intended for units with DIC
Unit SizeEA