SUPELCOSIL™ ABZ+Plus HPLC Column, 5 μm Particle Size
SUPELCOSIL™ ABZ+Plus HPLC Column, 5 μm Particle Size
Actual product may vary
SUPELCOSIL™ ABZ+Plus HPLC Column, 5 μm Particle Size
Offer both high deactivation and unique selectivity.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
5914810 cm21.2 mm
Catalog number 59148
Price (MYR)
-
Length (Metric):
10 cm
Diameter (Metric):
21.2 mm
SUPELCOSIL™ ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte's functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions. Suitable for L60 per USP.
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Specifications
Diameter (Metric)21.2 mm
Length (Metric)10 cm
Particle Size5 μm
Pore Size120 Å
Surface Area170 m2/g
Stationary PhaseAmide, Alkyl
Product LineSUPELCOSIL
MatrixAmide, Alkyl Phase/Silica Gel, Spherical Particle Platform
EndcappedYes
PhaseReversed Phase
Unit SizeEach