SUPELCOSIL™ LC-NH2-NP HPLC Column, 5 μm Particle Size
SUPELCOSIL™ LC-NH2-NP HPLC Column, 5 μm Particle Size
Actual product may vary
SUPELCOSIL™ LC-NH2-NP HPLC Column, 5 μm Particle Size
SUPELCOSIL™ LC-NH2-NP is an amino phase dedicated to normal phase chromatography. By employing special bonding technology, and avoiding water in manufacturing and testing the column, we have dramatically reduced the retention variation that is characteristic of normal phase chromatography.
Have Questions?
Catalog NumberLength (Metric)
5913225 cm
Catalog number 59132
Price (MYR)
-
Length (Metric):
25 cm
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Specifications
Diameter (Metric)4.6 mm
Length (Metric)25 cm
Particle Size5 μm
Pore Size120 Å
Surface Area170 m2/g
Stationary PhaseAmino (NH2)
Product LineSUPELCOSIL
MatrixAmino (NH2) Phase/Silica Gel, Spherical Particle Platform
EndcappedYes
PhaseReversed Phase
Unit SizeEach