Multidrop™ Combi Reagent Dispenser
Multidrop™ Combi Reagent Dispenser
Actual product may vary
Multidrop™ Combi Reagent Dispenser
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
5840340Multidrop Combi SMART+
5840330Multidrop Combi+
Catalog number 5840340
Price (MYR)
-
Description:
Multidrop Combi SMART+
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range
Multidrop Combi SMART:
  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability
Recommended for:
  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
Specifications
Accuracy
  • Small tube dispensing cassette: 2μL, ±10%; 10μL, ±5%
  • Standard tube dispensing cassette: 20μL, ±2%; 100μL, ±1%
Product TypeReagent Dispenser
DescriptionMultidrop Combi SMART+
Channels8
For Use With (Application)The instrument is intended for automated dispensing of up to eight different reagents simultaneously into a variety of microplates and strips in 96, 384 or 1536-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA.
For Use With (Equipment)Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and 6, 12, 24, 48, 96, or 384-well plate format. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm.
IncludesOne SMART+ standard tube dispensing cassette (cat. no. N22704)
InterfaceOn-board software and Serial RS-232 or USB for PC control
Plate Type6, 12, 24, 48, 96, and 384 well plates
Precision
  • Small tube dispensing cassette: 2 μL: CV ≤ 5%; 10 μL: CV ≤ 3% 
  • Standard tube dispensing cassette: 20 μL: CV ≤ 1.5%; 100 μL: ≤ CV 1% 
Speed
  • 96-well plate: 10μL in 5 sec.; 20μL in 5sec.; 100μL in 10 sec
  • 384-well plate: 1μL in 6 sec.; 5μL in 9 sec.; 10μL in 13 sec.; 20μL in 21 sec. 
  • 1536-well plate: 1μL in 15 sec.; 5μL in 28 sec. 
Volume (English)
  • Small tube dispensing cassette:  0.5-50μL, in 0.5μL increment 
  • Standard tube dispensing cassette:  5-2500μL, in 5 μL increment 
Volume (Metric) Dead<1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent) 
Warranty1 year
Dimensions (L x W x H)14 x 8.6 x 12.9 in. (35.5 x 22 x 33 cm)
Weight (English)20.1 lb.
Weight (Metric)9.1 kg
Electrical Requirements100 to 240 Vac 50/60Hz
Unit SizeEach
Frequently asked questions (FAQs)
Are the Multidrop Combi reagent dispensers compatible with Momentum Workflow Scheduling Software?
Yes, the Multidrop Combi reagent dispensers are compatible with the Momentum Software and can be integrated into automated workflow platforms. If you are planning to set up an automated workflow using the Momentum Software, you will also need a microplate mover such as the Orbitor RS2 Microplate Mover, Spinnaker Microplate Robot, or Spinnaker 3 Microplate Mover.
Do Multidrop Combi Reagent Dispensers (Cat. Nos. 58403xx and 58404xx) come with a laptop?
No, Multidrop Combi Reagent Dispensers are not sold with a laptop. This would need to be purchased separately. You can also use your own laptop or PC, as long as it meets the minimum PC requirements outlined in the Thermo Scientific FILLit Software for Multidrop Combi User Manual (pg 15).