ProPac™ SAX-10 HPLC Columns
ProPac™ SAX-10 HPLC Columns
Actual product may vary
ProPac™ SAX-10 HPLC Columns
Use ProPac™ SAX-10 LC Columns for exceptionally high resolution and efficiency when separating protein variants.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Column Format
54997250 mm4 mmAnalytical Column
Catalog number 54997
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Column Format:
Analytical Column
Use Thermo Scientific™ ProPac™ SAX-10 LC Columns for exceptionally high resolution and efficiency when separating protein variants. Based on a nonporous 10μm core particle that provides pressure stability, ProPac columns can resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Maximizing Resolution
  • ProPac SAX-10 LC column is a strong anion-exchanger with a quaternary amine functional group.
  • Column sizes range from 2mm I.D., for analytical work, up to 22mm I.D. for semiprep work.
Superior Performance for Protein Analysis
  • Ideal for characterization and quality control assessment of closely-related protein variants
  • Unequaled high-resolution separations
  • High-efficiency peaks with excellent recovery
  • Higher capacity than other columns based on nonporous particles
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Flow Rate1 mL/min
For Use WithLC
Length (Metric)250 mm
Max. Pressure3000 psi (207 bar)
Mobile Phase CompatibilitypH 2 to 12, 80% Compatible with Acetonitrile, Acetone, Methanol, Compatible with Nonionic and Zwitterionic Detergents, Incompatible with Anionic Detergents
Particle Size10 μm
pH2 to 12
Pore SizeNonporous
QuantityEach
Temperature60°C
USP TypeN/A
Stationary PhaseQuaternary Ammonium
Product LineProPac
Packing MaterialSpherical, Nonporous Polymer
Column TypeIon Exchange
PhaseIon Exchange
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEach